3-Brom-5-fluoranthranilsäure

Lieferant: FLUOROCHEM

Synonyme: 2-Amino-3-bromo-5-fluorobenzoic acid

034810-5G
FLUO034810-5GEA 104 EUR
FLUO034810-5G
3-Brom-5-fluoranthranilsäure
3-Brom-5-fluoranthranilsäure
Formel: C₇H₅BrFNO₂
Molecular Weight: 234,02 g/mol
MDL: MFCD00662509
CAS-Nummer: 259269-84-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR