Ethyl-5-amino-2-chlorbenzoat

Lieferant: FLUOROCHEM

Synonyme: Ethyl 5-amino-2-chlorobenzoate

034846-5G
FLUO034846-5GEA 347 EUR
FLUO034846-5G
Ethyl-5-amino-2-chlorbenzoat
Ethyl-5-amino-2-chlorbenzoat
Formel: C₉H₁₀ClNO₂
Molecular Weight: 199,6342 g/mol
MDL: MFCD00115822
CAS-Nummer: 64401-55-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR