1-Boc-3-bromoindole ≥97%
Lieferant: FLUOROCHEM
|
Achtung
|
Synonyme:
tert-Butyl 3-bromo-1H-indole-1-carboxylate, tert-Butyl-3-bromo-1H-indole-1-carboxylate, 3-Bromoindole-1-carboxylic acid tert-butyl ester
034920-5G
FLUO034920-5GEA
180
EUR
FLUO034920-5G
1-Boc-3-bromoindole ≥97%
1-Boc-3-bromoindole
Formel:
C₁₃H₁₄BrNO₂ Molecular Weight: 296,16 g/mol Siedepunkt: 367,9 °C (760 mmHg) |
MDL:
MFCD05864772 CAS-Nummer: 143259-56-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...