5-Brom-2-fluoranisol
Lieferant: FLUOROCHEM
Synonyme:
4-Bromo-1-fluoro-2-methoxybenzene
035061-25G
FLUO035061-25GEA
87.9
EUR
FLUO035061-25G
5-Brom-2-fluoranisol
5-Brom-2-fluoranisol
Formel:
C₇H₆BrFO Molecular Weight: 205.03 g/mol |
MDL:
MFCD01861130 CAS-Nummer: 103291-07-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...