2-Brom-6-fluor-4-nitroanilin

Lieferant: FLUOROCHEM

Synonyme: 2-Bromo-6-Fluoro-4-nitroaniline

035090-5G
FLUO035090-5GEA 288 EUR
FLUO035090-5G
2-Brom-6-fluor-4-nitroanilin
2-Brom-6-fluor-4-nitroanilin
Formel: C₆H₄BrFN₂O₂
Molecular Weight: 235,0100 g/mol
MDL: MFCD09800780
CAS-Nummer: 455-58-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR