5-Brom-3-chlor-2-iodbenzonitril
Lieferant: FLUOROCHEM
035129-1G
FLUO035129-1GEA
369
EUR
FLUO035129-1G
5-Brom-3-chlor-2-iodbenzonitril
5-Brom-3-chlor-2-iodbenzonitril
Formel:
C₇H₂BrClIN Molecular Weight: 342,359 g/mol |
MDL:
MFCD09800800 CAS-Nummer: 1000577-40-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...