3-Brom-2-fluor-6-iodbenzonitril

Lieferant: FLUOROCHEM

035141-5G
FLUO035141-5GEA 921 EUR
FLUO035141-5G
3-Brom-2-fluor-6-iodbenzonitril
3-Brom-2-fluor-6-iodbenzonitril
Formel: C₇H₂BrFIN
MDL: MFCD09800805
CAS-Nummer: 217816-66-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR