3-Azetidincarbonsäure
Lieferant: FLUOROCHEM
Synonyme:
3-Azetanecarboxylic acid, Azetidine-3-carboxylic acid
035210-25G
FLUO035210-25GEA
162
EUR
FLUO035210-25G
3-Azetidincarbonsäure
3-Azetidincarbonsäure
Formel:
C₄H₇NO₂ Molecular Weight: 101,11 g/mol |
MDL:
MFCD00191763 CAS-Nummer: 36476-78-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...