3,4-Dibromiodbenzol

Lieferant: FLUOROCHEM

035609-5G
FLUO035609-5GEA 347 EUR
FLUO035609-5G
3,4-Dibromiodbenzol
3,4-Dibromiodbenzol
Formel: C₆H₃Br₂I
MDL: MFCD09753707
CAS-Nummer: 909072-74-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR