2,4-Dibrom-3,6-difluoriodbenzol

Lieferant: FLUOROCHEM

035628-5G
FLUO035628-5GEA 549 EUR
FLUO035628-5G
2,4-Dibrom-3,6-difluoriodbenzol
2,4-Dibrom-3,6-difluoriodbenzol
Formel: C₆HBr₂F₂I
MDL: MFCD09878119
CAS-Nummer: 1000577-89-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR