1,4-Dibrom-3,5-difluorbenzol
Lieferant: FLUOROCHEM
Synonyme:
2,5-Dibromo-1,3-difluorobenzene
035631-5G
FLUO035631-5GEA
458
EUR
FLUO035631-5G
1,4-Dibrom-3,5-difluorbenzol
1,4-Dibromo-3,5-difluorobenzene
Formel:
C₆H₂Br₂F₂ Molecular Weight: 271,8849 g/mol |
MDL:
MFCD07783665 CAS-Nummer: 128259-71-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...