2,4-Dibrom-6-fluor-iodbenzol

Lieferant: FLUOROCHEM

Synonyme: 1,5-Dibromo-3-fluoro-2-iodobenzene

035640-25G
FLUO035640-25GEA 486 EUR
FLUO035640-25G
2,4-Dibrom-6-fluor-iodbenzol
2,4-Dibrom-6-fluor-iodbenzol
Formel: C₆H₂Br₂FI
Molecular Weight: 379,791 g/mol
MDL: MFCD00143072
CAS-Nummer: 202982-76-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR