4-Brom-2-chlor-5-fluoranilin

Lieferant: FLUOROCHEM

035677-5G
FLUO035677-5GEA 443 EUR
FLUO035677-5G
4-Brom-2-chlor-5-fluoranilin
4-Brom-2-chlor-5-fluoranilin
Formel: C₆H₄BrClFN
MDL: MFCD09878155
CAS-Nummer: 1000572-63-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR