4-Brom-2-chlor-5-fluor-1-iodbenzol

Lieferant: FLUOROCHEM

Synonyme: 4-Bromo-2-chloro-5-fluoroiodobenzene, 1-Bromo-5-chloro-2-fluoro-4-iodobenzene

035679-5G
FLUO035679-5GEA 723 EUR
FLUO035679-5G
4-Brom-2-chlor-5-fluor-1-iodbenzol
4-Brom-2-chlor-5-fluor-1-iodbenzol
Formel: C₆H₂BrClFI
Molecular Weight: 335.34 g/mol
MDL: MFCD09878157
CAS-Nummer: 1000572-73-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR