4-Brom-2-chlor-6-fluoranilin

Lieferant: FLUOROCHEM

035686-5G
FLUO035686-5GEA 237 EUR
FLUO035686-5G
4-Brom-2-chlor-6-fluoranilin
4-Brom-2-chlor-6-fluoranilin
Formel: C₆H₄BrClFN
Molecular Weight: 224,46 g/mol
MDL: MFCD09878163
CAS-Nummer: 885453-49-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR