5-Chlor-2-fluorphenylisothiocyanat

Lieferant: FLUOROCHEM

035696-5G
FLUO035696-5GEA 231 EUR
FLUO035696-5G
5-Chlor-2-fluorphenylisothiocyanat
5-Chlor-2-fluorphenylisothiocyanat
Formel: C₇H₃ClFNS
CAS-Nummer: 247170-25-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR