4-Brom-2,3-difluoranilin

Lieferant: FLUOROCHEM

035734-5G
FLUO035734-5GEA 171 EUR
FLUO035734-5G
4-Brom-2,3-difluoranilin
4-Brom-2,3-difluoranilin
Formel: C₆H₄BrF₂N
Molecular Weight: 208,0035 g/mol
MDL: MFCD09030643
CAS-Nummer: 112279-72-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR