4-(Difluormethyl)benzoesäure

Lieferant: FLUOROCHEM

035820-1G
FLUO035820-1GEA 365 EUR
FLUO035820-1G
4-(Difluormethyl)benzoesäure
4-(Difluoromethyl)benzoic acid
Formel: C₈H₆F₂O₂
Molecular Weight: 172,1289 g/mol
MDL: MFCD07784270
CAS-Nummer: 55805-21-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR