5-Brom-1,3-dichlor-2-fluorbenzol

Lieferant: FLUOROCHEM

Synonyme: 3,5-Dichloro-4-fluorobromobenzene

035887-25G
FLUO035887-25GEA 133 EUR
FLUO035887-25G
5-Brom-1,3-dichlor-2-fluorbenzol
5-Brom-1,3-dichlor-2-fluorbenzol
Formel: C₆H₂BrCl₂F
Molecular Weight: 243,8900 g/mol
MDL: MFCD09996901
CAS-Nummer: 17318-08-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR