Ethyl(4-fluorbenzoyl)formiat

Lieferant: FLUOROCHEM

Synonyme: Ethyl 2-(4-fluorophenyl)-2-oxoacetate

035925-5G
FLUO035925-5GEA 173 EUR
FLUO035925-5G
Ethyl(4-fluorbenzoyl)formiat
Ethyl(4-fluorbenzoyl)formiat
Formel: C₁₀H₉FO₃
Molecular Weight: 196.1751 g/mol
MDL: MFCD01319617
CAS-Nummer: 1813-94-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR