2-Nitroimidazol
Lieferant: FLUOROCHEM
|
Achtung
|
035942-5G
FLUO035942-5GEA
128
EUR
FLUO035942-5G
2-Nitroimidazol
2-Nitroimidazol
Formel:
C₃H₃N₃O₂ Schmelzpunkt: 279…284 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005185 CAS-Nummer: 527-73-1 EINECS: 208-425-5 UN: 2811 ADR: 6.1,III Merck Index: 13,00923 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...