3-Chlor-4-(trifluormethyl)benzylamin
Lieferant: FLUOROCHEM
036028-1G
FLUO036028-1GEA
131
EUR
FLUO036028-1G
3-Chlor-4-(trifluormethyl)benzylamin
3-Chlor-4-(trifluormethyl)benzylamin
Formel:
C₈H₇ClF₃N Molecular Weight: 209,60 g/mol |
MDL:
MFCD04972759 CAS-Nummer: 361393-93-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...