2-Fluor-m-kresol

Lieferant: FLUOROCHEM

036232-5G
FLUO036232-5GEA 159 EUR
FLUO036232-5G
2-Fluor-m-kresol
2-Fluor-m-kresol
Formel: C₇H₇FO
MDL: MFCD09835186
CAS-Nummer: 77772-72-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR