3,5-Difluor-4-hydroxybenzonitril

Lieferant: FLUOROCHEM

036238-1G
FLUO036238-1GEA 90.9 EUR
FLUO036238-1G
3,5-Difluor-4-hydroxybenzonitril
3,5-Difluor-4-hydroxybenzonitril
Formel: C₇H₃F₂NO
Molecular Weight: 155,1 g/mol
MDL: MFCD09033181
CAS-Nummer: 2967-54-6
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR