2-Amino-4-chlorpyrimidin

Lieferant: FLUOROCHEM

Synonyme: 4-chloropyrimidin-2-amine

036341-25G
FLUO036341-25GEA 282 EUR
FLUO036341-25G
2-Amino-4-chlorpyrimidin
2-Amino-4-chloropyrimidine
Formel: C₄H₄ClN₃
Molecular Weight: 129,55 g/mol
MDL: MFCD00038021
CAS-Nummer: 3993-78-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR