(5-Brom-2-pyridinyl)methanol
Lieferant: FLUOROCHEM
Synonyme:
5-Bromo-2-(hydroxymethyl)pyridine, 5-Bromo-2-hydroxymethylpyridine
036351-5G
FLUO036351-5GEA
110
EUR
FLUO036351-5G
(5-Brom-2-pyridinyl)methanol
(5-Brom-2-pyridinyl)methanol
Formel:
C₆H₆BrNO Molecular Weight: 188,0200 g/mol |
MDL:
MFCD04035597 CAS-Nummer: 88139-91-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...