4'-Amino-2-bromacetophenon
Lieferant: FLUOROCHEM
Synonyme:
4'-Aminophenacyl bromide
036701-5G
FLUO036701-5GEA
2930
EUR
FLUO036701-5G
4'-Amino-2-bromacetophenon
4'-Amino-2-bromacetophenon
Formel:
C₈H₈BrNO |
MDL:
MFCD09744997 CAS-Nummer: 23442-14-0 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...