2-Amino-4-bromphenol
Lieferant: FLUOROCHEM
Synonyme:
5-Brom-2-hydroxyanilin
036709-25G
FLUO036709-25GEA
119
EUR
FLUO036709-25G
2-Amino-4-bromphenol
2-Amino-4-bromphenol
Formel:
C₆H₆BrNO Molecular Weight: 188,02 g/mol |
MDL:
MFCD00235171 CAS-Nummer: 40925-68-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...