2-Amino-3,5-dibrompyrazin

Lieferant: FLUOROCHEM

Synonyme: Amino-3,5-dibromopyrazine

036760-25G
FLUO036760-25GEA 84.9 EUR
FLUO036760-25G
2-Amino-3,5-dibrompyrazin
2-Amino-3,5-dibromopyrazine
Formel: C₄H₃Br₂N₃
Molecular Weight: 252,90 g/mol
MDL: MFCD00673150
CAS-Nummer: 24241-18-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR