3-Brom-5-fluorpyridin
Lieferant: FLUOROCHEM
037143-1G
FLUO037143-1GEA
36.9
EUR
FLUO037143-1G
3-Brom-5-fluorpyridin
3-Bromo-5-fluoropyridine
Formel:
C₅H₃BrFN |
MDL:
MFCD04112555 CAS-Nummer: 407-20-5 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...