2,5-Difluor-m-toluylsäure
Lieferant: FLUOROCHEM
Synonyme:
2,5-Difluoro-3-methylbenzoic acid
037159-1G
FLUO037159-1GEA
659
EUR
FLUO037159-1G
2,5-Difluor-m-toluylsäure
2,5-Difluor-m-toluylsäure
Formel:
C₈H₆F₂O₂ Molecular Weight: 172,130 g/mol |
MDL:
MFCD09835286 CAS-Nummer: 1003710-01-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...