1-Chlor-3-fluor-2-nitrobenzol

Lieferant: FLUOROCHEM

037232-5G
FLUO037232-5GEA 120 EUR
FLUO037232-5G
1-Chlor-3-fluor-2-nitrobenzol
1-Chlor-3-fluor-2-nitrobenzol
Formel: C₆H₃ClFNO₂
MDL: MFCD06658264
CAS-Nummer: 64182-61-2
EINECS: 264-723-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR