Methyl-4-formylbenzoat
Lieferant: FLUOROCHEM
Synonyme:
Methyl terephthalaldehydate, Methyl 4-formylbenzoate
037428-25G
FLUO037428-25GEA
30.8
EUR
FLUO037428-25G
Methyl-4-formylbenzoat
Methyl-4-formylbenzoat
Formel:
C₉H₈O₃ Molecular Weight: 164,16 g/mol |
MDL:
MFCD00006950 CAS-Nummer: 1571-08-0 EINECS: 216-385-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...