5-Brom-2,4-dichloranisol
Lieferant: FLUOROCHEM
037538-25G
FLUO037538-25GEA
293
EUR
FLUO037538-25G
5-Brom-2,4-dichloranisol
5-Brom-2,4-dichloranisol
Formel:
C₇H₅BrCl₂O |
MDL:
MFCD11845926 CAS-Nummer: 174913-22-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...