4-Brom-2,3-difluorbenzoesäure

Lieferant: FLUOROCHEM

037604-5G
FLUO037604-5GEA 434 EUR
FLUO037604-5G
4-Brom-2,3-difluorbenzoesäure
4-Bromo-2,3-difluorobenzoic acid
Formel: C₇H₃BrF₂O₂
Molecular Weight: 236,9983 g/mol
MDL: MFCD07368121
CAS-Nummer: 194804-91-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR