3,5-Dibrombenzoesäure

Lieferant: FLUOROCHEM
Achtung

038167-25G
FLUO038167-25GEA 63.9 EUR
FLUO038167-25G
3,5-Dibrombenzoesäure
3,5-Dibromobenzoic acid
Formel: C₇H₄Br₂O₂
Molecular Weight: 279,92 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00051758
CAS-Nummer: 618-58-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR