Morpholin-3-on
Lieferant: FLUOROCHEM
Synonyme:
3-Morpholinone
038232-5G
FLUO038232-5GEA
55.4
EUR
FLUO038232-5G
Morpholin-3-on
Morpholin-3-on
Formel:
C₄H₇NO₂ |
MDL:
MFCD00631009 CAS-Nummer: 109-11-5 EINECS: 203-647-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...