5-Brom-2-(trifluormethyl)benzonitril

Lieferant: FLUOROCHEM

038249-5G
FLUO038249-5GEA 137 EUR
FLUO038249-5G
5-Brom-2-(trifluormethyl)benzonitril
5-Brom-2-(trifluormethyl)benzonitril
Formel: C₈H₃BrF₃N
Molecular Weight: 250,0153 g/mol
MDL: MFCD12026294
CAS-Nummer: 1208076-28-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR