3,6-Difluoro-2-methoxyphenol
Lieferant: FLUOROCHEM
Synonyme:
3,6-Difluoro-2-hydroxyanisole, 3,6-Difluoroguaiacol
038322-1G
FLUO038322-1GEA
1150
EUR
FLUO038322-1G
3,6-Difluoro-2-methoxyphenol
3,6-Difluoro-2-methoxyphenol
Formel:
C₇H₆F₂O₂ Molecular Weight: 160,118 g/mol |
MDL:
MFCD12827564 CAS-Nummer: 206198-07-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...