3-Brom-p-anisidin

Lieferant: FLUOROCHEM

Synonyme: 3-Bromo-4-methoxyaniline

038608-25G
FLUO038608-25GEA 162 EUR
FLUO038608-25G
3-Brom-p-anisidin
3-Brom-p-anisidin
Formel: C₇H₈BrNO
Molecular Weight: 202,0485 g/mol
MDL: MFCD04094339
CAS-Nummer: 19056-41-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR