3',5'-Difluoracetophenon

Lieferant: FLUOROCHEM

Synonyme: 1-(3,5-Difluorophenyl)ethan-1-one

038691-5G
FLUO038691-5GEA 764 EUR
FLUO038691-5G
3',5'-Difluoracetophenon
3',5'-Difluoracetophenon
Formel: C₈H₆F₂O
Molecular Weight: 156,13 g/mol
MDL: MFCD00042489
CAS-Nummer: 123577-99-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR