2-Methoxy-6-methylpyridin

Lieferant: FLUOROCHEM

040046-5G
FLUO040046-5GEA 30.6 EUR
FLUO040046-5G
2-Methoxy-6-methylpyridin
2-Methoxy-6-methylpyridine
Formel: C₇H₉NO
Molecular Weight: 123,16 g/mol
MDL: MFCD04112507
CAS-Nummer: 63071-03-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR