3-Methylisonicotinsäure

Lieferant: FLUOROCHEM

Synonyme: 3-Methyl-4-pyridinecarboxylic acid

040096-5G
FLUO040096-5GEA 344 EUR
FLUO040096-5G
3-Methylisonicotinsäure
3-Methylisonicotinsäure
Formel: C₇H₇NO₂
Molecular Weight: 137,1360 g/mol
MDL: MFCD00128870
CAS-Nummer: 4021-12-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR