2-Brom-3-fluoranilin

Lieferant: FLUOROCHEM

040426-25G
FLUO040426-25GEA 702 EUR
FLUO040426-25G
2-Brom-3-fluoranilin
2-Brom-3-fluoranilin
Formel: C₆H₅BrFN
Molecular Weight: 190.01 g/mol
MDL: MFCD07369915
CAS-Nummer: 111721-75-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR