2-Brom-4-nitroimidazol

Lieferant: FLUOROCHEM

Synonyme: 2-Brom--5-nitro1H-imidazol

040458-1G
FLUO040458-1GEA 31.1 EUR
FLUO040458-1G
2-Brom-4-nitroimidazol
2-Bromo-4-nitroimidazole
Formel: C₃H₂BrN₃O₂
Molecular Weight: 191,97 g/mol
MDL: MFCD09025308
CAS-Nummer: 65902-59-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR