Methyl-6-bromnicotinat

Lieferant: FLUOROCHEM

040655-5G
FLUO040655-5GEA 177 EUR
FLUO040655-5G
Methyl-6-bromnicotinat
Methyl-6-bromnicotinat
Formel: C₇H₆BrNO₂
MDL: MFCD04972371
CAS-Nummer: 26218-78-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR