1-Boc-piperazin

Lieferant: FLUOROCHEM

Synonyme: tert-Butyl 1-piperazinecarboxylate, tert-butyl piperazine-1-carboxylate

040783-5G
FLUO040783-5GEA 318 EUR
FLUO040783-5G
1-Boc-piperazin
1-Boc-piperazin
Formel: C₉H₁₈N₂O₂
Molecular Weight: 186,25 g/mol
MDL: MFCD00075265
CAS-Nummer: 57260-71-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR