3-Bromisatosäureanhydrid

Lieferant: FLUOROCHEM

Synonyme: 8-Bromo-1H-benzo[d][1,3]oxazine-2,4-dione

040876-1G
FLUO040876-1GEA 257 EUR
FLUO040876-1G
3-Bromisatosäureanhydrid
3-Bromisatosäureanhydrid
Formel: C₈H₄BrNO₃
Molecular Weight: 242,0300 g/mol
MDL: MFCD06738735
CAS-Nummer: 331646-98-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR