3-(Boc-amino)-3-(2-chlorphenyl)propionsäure
Lieferant: FLUOROCHEM
Synonyme:
3-((tert-Butoxycarbonyl)amino)-3-(2-chlorophenyl)propanoic acid
041173-10G
FLUO041173-10GEA
1160
EUR
FLUO041173-10G
3-(Boc-amino)-3-(2-chlorphenyl)propionsäure
3-(Boc-amino)-3-(2-chlorphenyl)propionsäure
Formel:
C₁₄H₁₈ClNO₄ Molecular Weight: 299,7500 g/mol |
MDL:
MFCD02090709 CAS-Nummer: 284493-66-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...