2,2,2,2'-Tetrafluoracetophenon

Lieferant: FLUOROCHEM

Synonyme: 2,2,2-Trifluoro-1-(2-fluorophenyl)ethanone

043106-1G
FLUO043106-1GEA 359 EUR
FLUO043106-1G
2,2,2,2'-Tetrafluoracetophenon
2,2,2,2'-Tetrafluoracetophenon
Formel: C₈H₄F₄O
Molecular Weight: 192,1104 g/mol
MDL: MFCD00040099
CAS-Nummer: 124004-75-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR